Learn More
ATCC Cervical Epithelial Cell Basal Medium, 485 mL
Supplier: ATCC PCS480032
Cervical Epithelial Cell Basal Medium is a sterile, phenol-red-free, liquid tissue culture medium that can be used as one component in a complete ATCC Primary Cell Solutions system. This serum-free system supports cervical epithelial cells derived from a patient's cervical tissue. Cervical Epithelial Cell Basal Medium contains essential and non-essential amino acids, vitamins, other organic compounds, trace minerals, and inorganic salts. To support the proliferation and plating efficiency of various types of cervical epithelial cells, Cervical Cell Basal Medium must be supplemented with the appropriate cell-specific growth kit. When using this complete media system, the growth of cervical epithelial cells is supported without feeder layers, extracellular matrix proteins, or other substrates.