Learn More
ATCC Cervical Epithelial Cell Growth Kit
Catalog No. 502383624
Encompass
Cervical Epithelial Cell Growth Kit contains components that when added to Cervical Epithelial Cell Basal Medium (ATCC PCS-480-032) create a complete ATCC Primary Cell Solutions culture environment for cervical epithelial cells derived from normal human cervical tissue (eg, Primary Cervical Epithelial Cells, Normal, Human, ATCC PCS-480-011). The serum-free medium supports normal cervical cell morphology, promote rapid growth, and proliferation. No feeder layers, extracellular matrix proteins, or other substrates are required.
Catalog No. 50-238-3624 Supplier ATCC Supplier No. PCS480042
Cervical Epithelial Cell Growth Kit contains components that when added to Cervical Epithelial Cell Basal Medium (ATCC PCS-480-032) create a complete ATCC Primary Cell Solutions culture environment for cervical epithelial cells derived from normal human cervical tissue (eg, Primary Cervical Epithelial Cells, Normal, Human, ATCC PCS-480-011). The serum-free medium supports normal cervical cell morphology, promote rapid growth, and proliferation. No feeder layers, extracellular matrix proteins, or other substrates are required.