Learn More
Aviva Systems Biology Corp CES1 Antibody - N-terminal region (ARP41877_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41877_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CES1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CES1. Alternate names: ACAT, CEH, CES2, HMSE, HMSE1, MGC117365, PCE-1, SES1, TGH, REHApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-58031