Learn More
Aviva Systems Biology Corp CES1 Antibody - C-terminal region (ARP41878_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41878_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CES1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CES1. Alternate names: CEH, CES2, HMSE, HMSE1, SES1, REH, TGH, ACAT, PCE-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58032