Learn More
Aviva Systems Biology Corp CES2 Antibody - middle region (ARP42733_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42733_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CES2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CES2. Alternate names: CE-2, CES2A1, PCE-2, iCEApplicationWB. Species reactivity: Human
Catalog No. 50-014-58320