Learn More
Aviva Systems Biology Corp CES2 Antibody - C-terminal region (ARP42158_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42158_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CES2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CES2. Alternate names: CE-2, iCE, PCE-2, CES2A1ApplicationWB, IHC. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-58127