Learn More
Aviva Systems Biology Corp CES3 Antibody - C-terminal region (ARP62006_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62006_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CES3. It was validated on Western Blot. . Alternate names: ES31, FLJ21736ApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-287-541