Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium, 99.8% (metals basis)

Catalog No. AA4300518
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4300518 50 g
AA4300503 1 g
AA4300506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4300518 Supplier Thermo Scientific Chemicals Supplier No. 04300518
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7440-46-2 | Cs | 132.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-46-2
Molecular Formula Cs
Molecular Weight (g/mol) 132.91
MDL Number MFCD00134037
InChI Key TVFDJXOCXUVLDH-UHFFFAOYSA-N
Synonym caesium, cesium-133, cesium, elemental, unii-1ksv9v4y4i, metals basis, 1ksv9v4y4i, cesio, zaesium, casium, zasium
PubChem CID 5354618
ChEBI CHEBI:30514
SMILES [Cs]

Specifications

Color Silver
Quantity 50 g
Assay Percent Range 99.8% (Metals basis)
IUPAC Name caesium
Chemical Name or Material Cesium
Hazard Category H260-H314-H361
Precautionary Statement P201-P202-P223-P231+P232-P260-P264b-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
DOTInformation DOT Class: 4.3, Packaging Group: I
EINECSNumber 231-155-4
RTECSNumber FK9225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.