Learn More
Strem, An Ascensus Company CESIUM ACETATE (99.9%-CS)-10G
SDP
Catalog No. 5090115745
Encompass
Cesium acetate; 99.9%-Cs; Lump; Strem Chemicals; 10g; Acetic acid, cesium salt; F.W. 191.95; C2H3CsO2; M.P. 194 deg.C; CAS: 3396-11-0
Catalog No. 50-901-15745 Supplier Strem, An Ascensus Company Supplier No. 93551010G
Cesium acetate; 99.9%-Cs; Lump; Strem Chemicals; 10g; Acetic acid, cesium salt; F.W. 191.95; C2H3CsO2; M.P. 194 deg.C; CAS: 3396-11-0