Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium Bicarbonate, MP Biomedicals

Catalog No. ICN210247 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN210247 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN210247 Supplier MP Biomedicals, Inc Supplier No. 0521024710

Chemical Identifiers

CAS 29703-01-3
Molecular Formula CHCsO3
Molecular Weight (g/mol) 193.92
MDL Number MFCD00010961
InChI Key ZMCUDHNSHCRDBT-UHFFFAOYSA-M
Synonym cesium bicarbonate, cesium hydrogen carbonate, caesium bicarbonate, caesium 1+ ion bicarbonate, cesium hydrogencarbonate, acmc-1cmhe, cho3.cs, caesium 1+ bicarbonate, carbonic acid, cesiumsalt 1:1
PubChem CID 2734983
IUPAC Name caesium(1+) hydrogen carbonate
SMILES [Cs+].OC([O-])=O

Specifications

Quantity 10 g
Formula Weight 193.92
Chemical Name or Material Cesium Bicarbonate
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.