Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium bromide, 99% (metals basis)

Catalog No. AA1349018
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1349018 50 g
AA1349030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1349018 Supplier Thermo Scientific Chemicals Supplier No. 01349018
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7787-69-1 | BrCs | 212.81 g/mol

Cesium bromide is used in optics as a beamsplitter component in wide-band spectrophotometers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-69-1
Molecular Formula BrCs
Molecular Weight (g/mol) 212.81
MDL Number MFCD00010954
InChI Key LYQFWZFBNBDLEO-UHFFFAOYSA-M
Synonym cesium bromide, caesium bromide, cesium bromide csbr, tricesium tribromide, csbr, unii-06m25edm3f, caesium 1+ ion bromide, cesiumbromide, br.cs, caesium i bromide
PubChem CID 24592
IUPAC Name cesium;bromide
SMILES [Br-].[Cs+]

Specifications

Boiling Point 1300°C
Melting Point 636°C
Color White
Physical Form Crystalline
Quantity 50 g
Odor Odorless
Refractive Index 1.6984
Sensitivity Hygroscopic
Assay Percent Range 99% (Metals basis)
Density 4.44 g/cm3 at 20°C
Solubility Information Very soluble in water. Soluble in alcohol. Insoluble in acetone
Formula Weight 212.81
Chemical Name or Material Cesium bromide
Show More Show Less
Hazard Category H302-H315-H319-H335-H361
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing and gloves. This material and/or its container must be disposed of as hazardous waste.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
EINECSNumber 232-130-0
RTECSNumber FK9275000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.