Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium bromide, ultra dry, 99.9% (metals basis)

Catalog No. aa1428909
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1428909 10 g
AA1428918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1428909 Supplier Thermo Scientific Chemicals Supplier No. 01428909
Only null left
Add to Cart

CAS: 7787-69-1 | BrCs | 212.81 g/mol

Cesium bromide is used as reagent, intermediate in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-69-1
Molecular Formula BrCs
Molecular Weight (g/mol) 212.81
MDL Number MFCD00010954
InChI Key LYQFWZFBNBDLEO-UHFFFAOYSA-M
Synonym cesium bromide, caesium bromide, cesium bromide csbr, tricesium tribromide, csbr, unii-06m25edm3f, caesium 1+ ion bromide, cesiumbromide, br.cs, caesium i bromide
PubChem CID 24592
IUPAC Name cesium;bromide
SMILES [Br-].[Cs+]

Specifications

Boiling Point 1300°C
Melting Point 636°C
Color White
Physical Form Beads
Quantity 10 g
Merck Index 14,2009
Solubility Information Soluble in water(55g/100mL at 20 C)
Formula Weight 212.81
Percent Purity 99.9%
Odor Odorless
Packaging Ampoule
Refractive Index 1.6984
Sensitivity Hygroscopic
Density 4.44 g/mL
Assay (metals basis)
Chemical Name or Material Cesium bromide, Ultra dry
Show More Show Less
Hazard Category H302-H315-H319-H335-H361
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
EINECSNumber 232-130-0
RTECSNumber FK9275000
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.