Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Cesium Carbonate
Spécifications
Spécifications
| CAS | 534-17-8 |
| Melting Point | 610°C |
| Molecular Formula | CCs2O3 |
| Linear Formula | Cs2CO3 |
| MDL Number | MFCD00010957 |
| Quantity | 5 g |
| Synonym | Carbonic acid dicesium |
| Molecular Weight (g/mol) | 325.82 g/mol |
missing translation for 'productTitle'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.
missing translation for 'spotOppurtunityImprovement'