Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cesium carbonate

Catalog No. 25564550G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-564-550G 50 g
25-564-510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25-564-550G Supplier Sigma Aldrich Supplier No. 25564550G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cesium carbonate

Specifications

Melting Point 610°C
CAS 534-17-8
Quantity 50 g
Molecular Formula CCs2O3
Linear Formula Cs2CO3
MDL Number MFCD00010957
Synonym Carbonic acid dicesium
Molecular Weight (g/mol) 325.82
Percent Purity 99.995% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.