Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium carbonate, Puratronic™, 99.994% (metals basis)

Catalog No. AA1211714
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 534-17-8 | CCs2O3 | 325.82 g/mol

Stainless Steel gauze is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 534-17-8
Molecular Formula CCs2O3
Molecular Weight (g/mol) 325.82
MDL Number MFCD00010957
InChI Key FJDQFPXHSGXQBY-UHFFFAOYSA-L
Synonym cesium carbonate, dicesium carbonate, caesium carbonate, cesiumcarbonate, carbonic acid, dicesium salt, cs2co3, unii-qqi20a14p4, cesium carbonate cs2co3, carbonic acid, cesium salt 1:2, dicaesium 1+ ion carbonate
PubChem CID 10796
SMILES [Cs+].[Cs+].[O-]C([O-])=O

Specifications

Melting Point 610°C (decomposition)
Color White
Physical Form Powder/Granules
Quantity 25 g
Assay Percent Range 99.994% (Metal basis)
Beilstein 4546405
Solubility Information Highly soluble in water. Soluble in alcohol,dimethyl sulfoxide and N,N-dimethylformamide. Insoluble in toluene and p-xylene.
IUPAC Name dicesium;carbonate
Formula Weight 325.82
Odor Odorless
Sensitivity Hygroscopic
Appearance White powder
Density 4.072
Chemical Name or Material Cesium carbonate
Show More Show Less
Hazard Category H318-H361f-H373
Precautionary Statement P201-P202-P260-P281-P305+P351+P338-P308+P313-P310-P501c
EINECSNumber 208-591-9
RTECSNumber FK9400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.