Learn More
Selleck Chemical LLC Cesium chloride S3642-1g
SDP
Catalog No. 503109264
Encompass_Preferred
Cesium chloride (CsCl) is the inorganic compound It is widely used medicine structure in isopycnic centrifugation for separating various types of DNA and is a reagent in analytical chemistry
Catalog No. 50-310-9264 Supplier Selleck Chemical LLC Supplier No. S36421G
Cesium chloride (CsCl) is the inorganic compound It is widely used medicine structure in isopycnic centrifugation for separating various types of DNA and is a reagent in analytical chemistry