Learn More
Research Products International Corp Cesium Chloride, 500 G
SDP
Catalog No. 502230262
Encompass_Preferred
Optical Density: 50% w/w solution in 1cm path cell at 260nm is <0.02.
Catalog No. 50-223-0262 Supplier Research Products International Corp Supplier No. C68050500.0
Optical Density: 50% w/w solution in 1cm path cell at 260nm is <0.02.