Learn More
Apexbio Technology LLC Cesium chloride 7647-17-8 50mg
SDP
Catalog No. 502240814
Encompass_Preferred
Cesium chloride (CAS 7647-17-8) is a small molecule known for its ability to block potassium channels specifically suppressing the pacemaker current (I sub f /sub ) and the hyperpolarization-activated cationic current (I sub h /sub ) In neuronal models it has been shown to inhibit activation of caspase-3 and prevent apoptosis in cerebellar granule neurons subjected to serum and potassium deprivation a process associated with inactivation of GSK-3 Cesium chloride is widely employed in neurophysiological studies to investigate ion channel function and the molecular mechanisms underlying neuronal survival
This item is not returnable. View return policy
Catalog No. 50-224-0814 Supplier Apexbio Technology LLC Supplier No. A446850
Cesium chloride (CAS 7647-17-8) is a small molecule known for its ability to block potassium channels specifically suppressing the pacemaker current (I sub f /sub ) and the hyperpolarization-activated cationic current (I sub h /sub ) In neuronal models it has been shown to inhibit activation of caspase-3 and prevent apoptosis in cerebellar granule neurons subjected to serum and potassium deprivation a process associated with inactivation of GSK-3 Cesium chloride is widely employed in neurophysiological studies to investigate ion channel function and the molecular mechanisms underlying neuronal survival