Learn More
Apexbio Technology LLC Cesium chloride 7647-17-8 10mM (in 1mL H2O)
SDP
Catalog No. 5000094795
Encompass_Preferred
Cesium chloride (CAS 7647-17-8) is a small molecule known for its ability to block potassium channels specifically suppressing the pacemaker current (I sub f /sub ) and the hyperpolarization-activated cationic current (I sub h /sub ) In neuronal models it has been shown to inhibit activation of caspase-3 and prevent apoptosis in cerebellar granule neurons subjected to serum and potassium deprivation a process associated with inactivation of GSK-3 Cesium chloride is widely employed in neurophysiological studies to investigate ion channel function and the molecular mechanisms underlying neuronal survival
Catalog No. 50-000-94795 Supplier Apexbio Technology LLC Supplier No. A44685.1
Cesium chloride (CAS 7647-17-8) is a small molecule known for its ability to block potassium channels specifically suppressing the pacemaker current (I sub f /sub ) and the hyperpolarization-activated cationic current (I sub h /sub ) In neuronal models it has been shown to inhibit activation of caspase-3 and prevent apoptosis in cerebellar granule neurons subjected to serum and potassium deprivation a process associated with inactivation of GSK-3 Cesium chloride is widely employed in neurophysiological studies to investigate ion channel function and the molecular mechanisms underlying neuronal survival