Learn More
Medchemexpress LLC Cesium chloride | 7647-17-8 | MFCD00010955 | 99.9% | 168.36 g/mol | CsCl | 10 G
SDP
Catalog No. 5000242744
Encompass_Preferred
Cesium chloride is an inorganic salt (CsCl) used as a reagent and a potassium channel blocker in biochemical and physiological assays. Supplied as a dry, high-purity crystalline powder, it is suitable for molecular biology and analytical workflows; store at room temperature and keep dry.
  • High assay purity (≈99.9%) for reproducible results.
  • Available in multiple pack sizes for flexible use.
  • Crystalline powder form that dissolves readily in water.
  • Used as a potassium channel blocker in electrophysiology assays.
  • Stable at room temperature when kept dry and sealed.
Catalog No. 50-002-42744 Supplier Medchemexpress LLC Supplier No. HY10775410G
Cesium chloride is an inorganic salt (CsCl) used as a reagent and a potassium channel blocker in biochemical and physiological assays. Supplied as a dry, high-purity crystalline powder, it is suitable for molecular biology and analytical workflows; store at room temperature and keep dry.
  • High assay purity (≈99.9%) for reproducible results.
  • Available in multiple pack sizes for flexible use.
  • Crystalline powder form that dissolves readily in water.
  • Used as a potassium channel blocker in electrophysiology assays.
  • Stable at room temperature when kept dry and sealed.