Learn More
Medchemexpress LLC Cesium chloride | 7647-17-8 | MFCD00010955 | 99.9% | 168.36 g·mol⁻¹ | CsCl | 25 G
SDP
Catalog No. 5000240323
Encompass_Preferred
Cesium chloride (CsCl) is an inorganic salt used in biochemical research as a potassium channel blocker and as a reagent for density gradient centrifugation. It is provided as a high-purity laboratory reagent suitable for electrophysiology and molecular biology applications; consult the certificate of analysis for batch-specific purity and handling information.
  • High purity (99.9%) suitable for analytical and research use.
  • Functions as a potassium channel blocker in electrophysiology studies.
  • Applicable for density gradient centrifugation in molecular biology.
  • Supplied in laboratory pack sizes for convenient handling.
  • Certificate of analysis available for batch-specific specifications.
Catalog No. 50-002-40323 Supplier Medchemexpress LLC Supplier No. HY10775425G
Cesium chloride (CsCl) is an inorganic salt used in biochemical research as a potassium channel blocker and as a reagent for density gradient centrifugation. It is provided as a high-purity laboratory reagent suitable for electrophysiology and molecular biology applications; consult the certificate of analysis for batch-specific purity and handling information.
  • High purity (99.9%) suitable for analytical and research use.
  • Functions as a potassium channel blocker in electrophysiology studies.
  • Applicable for density gradient centrifugation in molecular biology.
  • Supplied in laboratory pack sizes for convenient handling.
  • Certificate of analysis available for batch-specific specifications.