Learn More
Medchemexpress LLC Cesium chloride | 7647-17-8 | MFCD00010955 | 99.9% | 168.36 g/mol | CsCl | 50 G
SDP
Catalog No. 5000241178
Encompass_Preferred
Cesium chloride (CsCl) is an inorganic salt widely used in biochemical and molecular biology applications as a density gradient medium and as a potassium channel blocker. It is supplied as a high-purity, crystalline reagent that is readily soluble in water and suitable for laboratory use.
  • High purity for laboratory applications.
  • Suitable for density gradient centrifugation and nucleic acid separation.
  • Acts as a potassium channel blocker in physiological studies.
  • Water-soluble crystalline salt.
  • Available in multiple laboratory-scale pack sizes.
Catalog No. 50-002-41178 Supplier Medchemexpress LLC Supplier No. HY10775450G
Cesium chloride (CsCl) is an inorganic salt widely used in biochemical and molecular biology applications as a density gradient medium and as a potassium channel blocker. It is supplied as a high-purity, crystalline reagent that is readily soluble in water and suitable for laboratory use.
  • High purity for laboratory applications.
  • Suitable for density gradient centrifugation and nucleic acid separation.
  • Acts as a potassium channel blocker in physiological studies.
  • Water-soluble crystalline salt.
  • Available in multiple laboratory-scale pack sizes.