Learn More
Specifications
Specifications
| CAS | 7647-17-8 |
| Quantity | 6 x 100 g |
| Molecular Formula | ClCs |
| MDL Number | MFCD00010955 |
| Synonym | cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride |
| InChI Key | AIYUHDOJVYHVIT-UHFFFAOYSA-M |
| SMILES | [Cl-].[Cs+] |
| IUPAC Name | caesium(1+) chloride |
| Molecular Weight (g/mol) | 168.36 |
| PubChem CID | 24293 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.