Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium Chloride Lanthanum Chloride Buffer, Solstice

Catalog No. 60048401 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-401 250 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-048-401 Supplier Solstice Supplier No. 95164250ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TraceSELECT™, for flame atomic absorption spectrometry

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cesium chloride Lanthanum chloride buffer
pH <1
Quantity 250 mL
MDL Number MFCD00152971
UN Number UN3082
Solubility Information Soluble in Water
Packaging Poly Bottle
Product Line TraceSELECT™
Appearance Violet Liquid
Density 1.17g/mL at 20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.