Learn More
Research Products International Corp Cesium Chloride, Optical Grade, 100 Grams
SDP
Catalog No. 50213259
Encompass_Preferred
Optical Density: 50% w/w solution in 1cm path cell at 260nm is <0.02.
Catalog No. 50-213-259 Supplier Research Products International Corp Supplier No. C68050100.0
Optical Density: 50% w/w solution in 1cm path cell at 260nm is <0.02.