Learn More
Alkali Scientific Cesium Chloride, Optical Grade, 1 Kg
SDP
Catalog No. 5000660720
Encompass_Preferred
Cesium Chloride Optical Grade, supplied in a 1 kg pack, is a specialized reagent used for creating density gradients in molecular biology and chemistry applications. This high-purity salt is essential for the separation of nucleic acids, proteins, and other biomolecules based on their density. Cesium Chloride is commonly used in ultracentrifugation processes to isolate and purify samples for analysis. The 1 kg size is perfect for laboratories that require large volumes of Cesium Chloride for routine applications such as DNA or RNA separations, protein purification, and other biochemical procedures that depend on precise density gradients.
Catalog No. 50-006-60720 Supplier Alkali Scientific Supplier No. CES002CP
Cesium Chloride Optical Grade, supplied in a 1 kg pack, is a specialized reagent used for creating density gradients in molecular biology and chemistry applications. This high-purity salt is essential for the separation of nucleic acids, proteins, and other biomolecules based on their density. Cesium Chloride is commonly used in ultracentrifugation processes to isolate and purify samples for analysis. The 1 kg size is perfect for laboratories that require large volumes of Cesium Chloride for routine applications such as DNA or RNA separations, protein purification, and other biochemical procedures that depend on precise density gradients.