Learn More
Strem, An Ascensus Company CAS# 13400-13-0. 150G. Cesium fluoride (99.9%-Cs). MFCD00010960
SDP
Catalog No. 501963603
Encompass
CAS# 13400-13-0. 50G. Cesium fluoride (99.9%-Cs). MFCD00010960. Molecular Formula: CsF. Molecular Weight: 151.90. Color/Form: white pwdr. Stability: hygroscopic. Strem# 93-5518. www.strem.com/catalog/v3/93-5518/
Catalog No. 50-196-3603 Supplier Strem, An Ascensus Company Supplier No. 93551850G
CAS# 13400-13-0. 50G. Cesium fluoride (99.9%-Cs). MFCD00010960. Molecular Formula: CsF. Molecular Weight: 151.90. Color/Form: white pwdr. Stability: hygroscopic. Strem# 93-5518. www.strem.com/catalog/v3/93-5518/