Learn More
Strem, An Ascensus Company CESIUM FLUORIDE (99.9%-CS)-10G
SDP
Catalog No. 5090115752
Encompass
Cesium fluoride; 99.9%-Cs; White powder; Strem Chemicals; 10g; Cesium monofluoride; F.W. 151.90; CsF; M.P. 682 deg.C; B.P. 1251 deg.C; CAS: 13400-13-0
Catalog No. 50-901-15752 Supplier Strem, An Ascensus Company Supplier No. 93551810G
Cesium fluoride; 99.9%-Cs; White powder; Strem Chemicals; 10g; Cesium monofluoride; F.W. 151.90; CsF; M.P. 682 deg.C; B.P. 1251 deg.C; CAS: 13400-13-0