Learn More
Strem, An Ascensus Company CAS# 13400-13-0. 25g. Cesium fluoride (99+%-Cs). MFCD00010960
SDP
Catalog No. 5090115754
Encompass
CAS# 13400-13-0. 25g. Cesium fluoride (99+%-Cs). MFCD00010960. Molecular Weight: 151.90. Molecular Formula: CsF. Color/form: white pwdr. Strem# 93-5519. http://www.strem.com/catalog/v/93-5519/
Catalog No. 50-901-15754 Supplier Strem, An Ascensus Company Supplier No. 93551925G
CAS# 13400-13-0. 25g. Cesium fluoride (99+%-Cs). MFCD00010960. Molecular Weight: 151.90. Molecular Formula: CsF. Color/form: white pwdr. Strem# 93-5519. http://www.strem.com/catalog/v/93-5519/