Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium Green 85.0+%, TCI America™
SDP

Catalog No. C280650MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C280650MG 50 mg
C2806250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C280650MG Supplier TCI America Supplier No. C280650MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1611477-45-2
Molecular Formula C32H33NO8
Molecular Weight (g/mol) 559.615
InChI Key HCDBZEWQACQBAN-UHFFFAOYSA-N
PubChem CID 124523977
IUPAC Name 4-[4-[4-[2-[2-[2-[2-(4-nitrophenoxy)ethoxy]ethoxy]ethoxy]ethoxy]phenyl]phenyl]phenol
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)O)C3=CC=C(C=C3)OCCOCCOCCOCCOC4=CC=C(C=C4)[N+](=O)[O-]

Specifications

Synonym 4′C′C-[2-[2-[2-[2-(4-Nitrophenoxy)ethoxy]ethoxy]ethoxy]ethoxy]-[1,1′C:4′C,1′C′C-terphenyl]-4-ol
Quantity 50 mg
Physical Form Crystal/Powder
Formula Weight 559.62
Percent Purity ≥85.0% (HPLC)
Grade Research
Chemical Name or Material Cesium Green
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.