Learn More
Strem, An Ascensus Company CAS# 12260-45-6. 10g. Cesium hydroxide, hydrate (99.9%-Cs). MFCD00149664
SDP
Supplier:  Strem, An Ascensus Company 93552110G
Encompass
CAS# 12260-45-6. 10g. Cesium hydroxide, hydrate (99.9%-Cs). MFCD00149664. Molecular Weight: 149.91. Molecular Formula: CsOH∙XH2O. Color/form: xtl. (15-20% H2O) Strem# 93-5521. http://www.strem.com/catalog/v/93-5521/
Catalog No. 50-901-15758