Learn More
CTL Scientific Supply Corp CESIUM IODIDE 50 GM
SDP
Catalog No. 50300423 Shop All CTL Scientific Supply Products
Encompass
Ionic compound often used as the input phosphor of an x-ray image intensifier tube found in fluoroscopy equipment. 50 gm bottle
Catalog No. 50-300-423 Supplier CTL Scientific Supply Corp Supplier No. 695300
Ionic compound often used as the input phosphor of an x-ray image intensifier tube found in fluoroscopy equipment. 50 gm bottle