Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cesium iodide | 7789-17-5 | MFCD00010962 | 259.8099 g/mol | CsI | 10 G
SDP

Catalog No. 5000209648
Encompass_Preferred

Cesium iodide (CsI) is an inorganic ionic compound commonly used in optics and radiation detection. It is a white crystalline solid used as a scintillator, detector window material, and analytical standard in research.

  • Chemical formula: CsI.
  • Molecular weight: 259.8099 g/mol.
  • CAS number: 7789-17-5.
  • MDL/MFCD number: MFCD00010962.
  • Appearance: White crystalline solid.
  • Typical package size: 10 G.
  • Common applications: scintillation detectors, optical windows, analytical standards.

Catalog No. 50-002-09648 Supplier Medchemexpress LLC Supplier No. HYW04016010G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Cesium iodide (CsI) is an inorganic ionic compound commonly used in optics and radiation detection. It is a white crystalline solid used as a scintillator, detector window material, and analytical standard in research.

  • Chemical formula: CsI.
  • Molecular weight: 259.8099 g/mol.
  • CAS number: 7789-17-5.
  • MDL/MFCD number: MFCD00010962.
  • Appearance: White crystalline solid.
  • Typical package size: 10 G.
  • Common applications: scintillation detectors, optical windows, analytical standards.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.