Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cesium iodide | 7789-17-5 | MFCD00010962 | 259.81 g·mol⁻¹ | CsI | 50 G
SDP

Catalog No. 5000209102
Encompass_Preferred

Cesium iodide (CsI) is an inorganic ionic salt used in analytical and materials research. It is a white crystalline solid with applications in optics, detectors, and laboratory analysis.

  • Inorganic cesium salt used in research applications.
  • Molecular formula CsI; molecular weight 259.81 g·mol⁻¹.
  • CAS number 7789-17-5.
  • Supplied as a solid; store sealed at 4°C and protect from moisture.
  • Trace metal content is reported in certificates of analysis (ppm levels).

Catalog No. 50-002-09102 Supplier Medchemexpress LLC Supplier No. HYW04016050G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Cesium iodide (CsI) is an inorganic ionic salt used in analytical and materials research. It is a white crystalline solid with applications in optics, detectors, and laboratory analysis.

  • Inorganic cesium salt used in research applications.
  • Molecular formula CsI; molecular weight 259.81 g·mol⁻¹.
  • CAS number 7789-17-5.
  • Supplied as a solid; store sealed at 4°C and protect from moisture.
  • Trace metal content is reported in certificates of analysis (ppm levels).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.