Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cesium iodide | 7789-17-5 | MFCD00010962 | 99.9% | 259.81 g/mol | CsI | 25 G
SDP

Catalog No. 5000209529
Encompass_Preferred

Cesium iodide (CsI) is an inorganic cesium salt supplied as a white to off-white crystalline solid for laboratory and analytical use. It is identified by CAS 7789-17-5 and has a molecular weight of 259.81 g/mol. The manufacturer certificate of analysis reports trace metal impurities ≤100 ppm.

  • Chemical formula: CsI
  • Molecular weight: 259.81 g/mol
  • Cas number: 7789-17-5
  • Physical description: white to off-white crystals
  • Purity: 99.9% (typical, trace metals basis)
  • Trace metals: ≤100 ppm (per COA)
  • Pack size: 25 G

Catalog No. 50-002-09529 Supplier Medchemexpress LLC Supplier No. 25GHYW040160
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cesium iodide (CsI) is an inorganic cesium salt supplied as a white to off-white crystalline solid for laboratory and analytical use. It is identified by CAS 7789-17-5 and has a molecular weight of 259.81 g/mol. The manufacturer certificate of analysis reports trace metal impurities ≤100 ppm.

  • Chemical formula: CsI
  • Molecular weight: 259.81 g/mol
  • Cas number: 7789-17-5
  • Physical description: white to off-white crystals
  • Purity: 99.9% (typical, trace metals basis)
  • Trace metals: ≤100 ppm (per COA)
  • Pack size: 25 G

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.