Learn More
Sigma Aldrich Fine Chemicals Biosciences Cesium methanesulfonate us25G
Catalog No. 502903527
Encompass
Cesium methanesulfonate is used as a component of intracellular recording SOLUTIONons of electrodes for patch clamp techniques in cell electrophysiology studies. Cesium methanesulfonate is used as an current carrier as cesium acts as a potassium (K) channel blocker while methanesulfonate is not transported through chloride (Cl-) channels.
Catalog No. 50-290-3527 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C142625G
Cesium methanesulfonate is used as a component of intracellular recording SOLUTIONons of electrodes for patch clamp techniques in cell electrophysiology studies. Cesium methanesulfonate is used as an current carrier as cesium acts as a potassium (K) channel blocker while methanesulfonate is not transported through chloride (Cl-) channels.