Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cesium nitrate

Catalog No. 20215050G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-215-050G 50 g
20-215-010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 20-215-050G Supplier Sigma Aldrich Supplier No. 20215050G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cesium nitrate

Specifications

Melting Point 414°C
CAS 7789-18-6
Quantity 50 g
Molecular Formula CsNO3
Linear Formula CsNO3
MDL Number MFCD00010963
Synonym Nitric acid cesium salt
Molecular Weight (g/mol) 194.91
Percent Purity 99.99% trace metals basis
Density 3.685 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.