Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium sulfate, 99.9% (metals basis)

Catalog No. p-4407373
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1455914 25 g
AA1455922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1455914 Supplier Thermo Scientific Chemicals Supplier No. 01455914
Only null left
Add to Cart

CAS: 10294-54-9 | Cs2O4S | 361.87 g/mol

Cesium sulfate is used to prepare dense aqueous solutions, which finds application in isopycnic centrifugation especially for DNA purification and recovery. It acts as a catalyst for sulfur dioxide oxidation with vanadium or vanadium pentoxide. Further, it acts as a metal oxide catalyst enhancer for heterogeneous processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-54-9
Molecular Formula Cs2O4S
Molecular Weight (g/mol) 361.87
MDL Number MFCD00010959
InChI Key FLJPGEWQYJVDPF-UHFFFAOYSA-L
Synonym cesium sulfate, dicesium sulfate, sulfuric acid, dicesium salt, caesium sulphate, caesium sulfate, unii-8d6r91cs62, dicaesium 1+ ion sulfate, cesiumsulfate, dicaesium 1+ sulfate
PubChem CID 25137
SMILES [Cs+].[Cs+].[O-]S([O-])(=O)=O

Specifications

Melting Point 1010°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Soluble in water. Insoluble in ethanol and acetone.
IUPAC Name dicaesium(1+) sulfate
Formula Weight 361.87
Sensitivity Hygroscopic
Appearance White powder
Density 4.243
Chemical Name or Material Cesium sulfate
Show More Show Less
Hazard Category H302-H361f-H373
Precautionary Statement P201-P202-P260-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
EINECSNumber 233-662-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.