Learn More
FUJIFILM Biosciences Cesium Trifluoroacetate Solu
Catalog No. 5000450303
Encompass_Preferred
Cesium trifluoroacetate is a new medium for equilibrium density gradient centrifugation replacing cesium chloride Its recovery rate is stable and is particularly suited to isolation of highly pure and intact RNAs It is characterized by the inhibitory effect on RNase activity and is effective for purification of mRNAs It also solubilizes proteins and accelerates dissociation of proteins from nucleic acids and therefore it can isolate highly pure RNAs withuot protein contamination at high yields For research use only RUO
Catalog No. 50-004-50303 Supplier FUJIFILM Biosciences Supplier No. 03113681
Cesium trifluoroacetate is a new medium for equilibrium density gradient centrifugation replacing cesium chloride Its recovery rate is stable and is particularly suited to isolation of highly pure and intact RNAs It is characterized by the inhibitory effect on RNase activity and is effective for purification of mRNAs It also solubilizes proteins and accelerates dissociation of proteins from nucleic acids and therefore it can isolate highly pure RNAs withuot protein contamination at high yields For research use only RUO