Learn More
ATCC CESS; Leukemia; Human (Homo sapiens)
Catalog No. 502383203
Encompass
CESS is a lymphoblast cell isolated from the blood of a White male with myelomonocytic leukemia.
Catalog No. 50-238-3203 Supplier ATCC Supplier No. TIB190
CESS is a lymphoblast cell isolated from the blood of a White male with myelomonocytic leukemia.