Learn More
Med Vet International Zoetis, Cestex Tablets 25mg, 50 Tabs
Catalog No. 502828333
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Cestex (epsiprantel) tablets are indicated for the removal of tapeworms in cats (Dipylidium caninum and Taenia taeniaeformis) and dogs (Dipylidium caninum and Taenia pisiformis). Only one dose is needed. Fasting is not necessary or recommended. Do not use in animals younger than 7 weeks
Catalog No. 50-282-8333 Supplier Med Vet International Supplier No. RXCESTEX2550
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Cestex (epsiprantel) tablets are indicated for the removal of tapeworms in cats (Dipylidium caninum and Taenia taeniaeformis) and dogs (Dipylidium caninum and Taenia pisiformis). Only one dose is needed. Fasting is not necessary or recommended. Do not use in animals younger than 7 weeks