Learn More
Med Vet International Zoetis, Cestex, Oragne, 100mg, 50 Tablets
Catalog No. 502828331
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Cestex (epsiprantel) tablets are indicated for the removal of tapeworms in cats (Dipylidium caninum and Taenia taeniaeformis) and dogs (Dipylidium caninum and Taenia pisiformis). Only one dose is needed. Fasting is not necessary or recommended. Do not use in animals younger than 7 weeks.
Catalog No. 50-282-8331 Supplier Med Vet International Supplier No. RXCESTEX10050
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Cestex (epsiprantel) tablets are indicated for the removal of tapeworms in cats (Dipylidium caninum and Taenia taeniaeformis) and dogs (Dipylidium caninum and Taenia pisiformis). Only one dose is needed. Fasting is not necessary or recommended. Do not use in animals younger than 7 weeks.