Learn More
CAPTIVATE BIO LLC CET Small Molecules for 20L
Catalog No. 5000876356
Encompass_Preferred
Captivate Bio s newly optimized CET Cocktail Kits are designed to enhance cell survival and promote growth of genetically stable hPSCs Kits are configured to address specific workflows Three versions include CET Spark for 1L CET Ignite for 5L and CET Titan for 20L or more
Catalog No. 50-008-76356 Supplier CAPTIVATE BIO LLC Supplier No. CET03B
Captivate Bio s newly optimized CET Cocktail Kits are designed to enhance cell survival and promote growth of genetically stable hPSCs Kits are configured to address specific workflows Three versions include CET Spark for 1L CET Ignite for 5L and CET Titan for 20L or more