Learn More
Medchemexpress LLC HY-119964 5mg Medchemexpress, Cetaben CAS:55986-43-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1199645MG
Encompass_Preferred
Medchemexpress, HY-119964 5mg Cetaben CAS:55986-43-1 Cetaben is a PPARα-independent peroxisome proliferator. Cetaben is a non-fibrate hypolipidemic drug and potently reduces the concentration of cholesterol and triglycerides. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9561