Learn More
Medchemexpress LLC Cetalkonium chloride | 122-18-9 | 99.4% | 1 G
SDP
Catalog No. 5000413174
Encompass_Preferred
Cetalkonium chloride is an ammonium antiseptic agent used in many topical agents for infections of the mouth, throat, and eye. It acts as an anti-inflammatory amphiphilic agent and a cationic surfactant that improves the precorneal residence time and bioavailability of drugs. It interacts with negative charges on the bacterial cell membrane, disrupting its integrity, causing leakage of cell contents, and ultimately leading to bacterial death.
  • Ammonium antiseptic agent
  • Anti-inflammatory amphiphilic agent
  • Cationic surfactant
  • Exhibits antibacterial activity against Gram-positive bacteria
  • Increases healing speed in scraped human corneal epithelial cells
  • Promotes corneal wound healing and has anti-inflammatory properties
  • Used in topical agents for infections of the mouth, throat, and eye
  • Improves the precorneal residence time and bioavailability of drugs
Catalog No. 50-004-13174 Supplier Medchemexpress LLC Supplier No. HYB15971G
Cetalkonium chloride is an ammonium antiseptic agent used in many topical agents for infections of the mouth, throat, and eye. It acts as an anti-inflammatory amphiphilic agent and a cationic surfactant that improves the precorneal residence time and bioavailability of drugs. It interacts with negative charges on the bacterial cell membrane, disrupting its integrity, causing leakage of cell contents, and ultimately leading to bacterial death.
  • Ammonium antiseptic agent
  • Anti-inflammatory amphiphilic agent
  • Cationic surfactant
  • Exhibits antibacterial activity against Gram-positive bacteria
  • Increases healing speed in scraped human corneal epithelial cells
  • Promotes corneal wound healing and has anti-inflammatory properties
  • Used in topical agents for infections of the mouth, throat, and eye
  • Improves the precorneal residence time and bioavailability of drugs