Learn More
Medchemexpress LLC Cetearyl alcohol | 67762-27-0 | 99.1% | 512.93 g/mol | C34H72O2 | 100 G
SDP
Catalog No. 5000281058
Encompass_Preferred
Cetearyl alcohol is a pharmaceutical-grade fatty alcohol commonly used as an excipient and emollient in topical and cosmetic formulations. It is supplied as a white to off-white solid and provides emulsification, thickening, and moisturizing properties that improve texture and stability in creams and lotions.
  • High purity suitable for formulation and research use.
  • Solid, white to off-white form for convenient handling.
  • Acts as an emulsifier and stabilizer in oil-in-water systems.
  • Provides emollient and thickening properties to improve product texture.
  • Available in lab-scale packaging including 100 G.
  • Certificate of analysis and safety data sheet available for quality and safety assurance.
Catalog No. 50-002-81058 Supplier Medchemexpress LLC Supplier No. HYW422419100G
Cetearyl alcohol is a pharmaceutical-grade fatty alcohol commonly used as an excipient and emollient in topical and cosmetic formulations. It is supplied as a white to off-white solid and provides emulsification, thickening, and moisturizing properties that improve texture and stability in creams and lotions.
  • High purity suitable for formulation and research use.
  • Solid, white to off-white form for convenient handling.
  • Acts as an emulsifier and stabilizer in oil-in-water systems.
  • Provides emollient and thickening properties to improve product texture.
  • Available in lab-scale packaging including 100 G.
  • Certificate of analysis and safety data sheet available for quality and safety assurance.