Learn More
Medchemexpress LLC Cetearyl alcohol | 67762-27-0 | MFCD01076350 | 99.1% | 512.94 g/mol | C34H72O2 | 500 G
SDP
Catalog No. 5000281163
Encompass_Preferred
Cetearyl alcohol is a mixed C16-C18 fatty alcohol used as an emollient, thickener, and nonionic emulsifier in pharmaceutical and cosmetic formulations. Supplied as a white, waxy solid, it improves texture, stability, and spreadability of creams, lotions, and ointments.
  • Waxy white solid at room temperature.
  • Acts as an emollient and thickening agent.
  • Functions as a nonionic emulsifier to stabilize oil-in-water systems.
  • Improves texture, spreadability, and product stability.
  • Suitable for topical pharmaceutical and cosmetic formulations.
  • High purity (99.1%).
Catalog No. 50-002-81163 Supplier Medchemexpress LLC Supplier No. HYW422419500G
Cetearyl alcohol is a mixed C16-C18 fatty alcohol used as an emollient, thickener, and nonionic emulsifier in pharmaceutical and cosmetic formulations. Supplied as a white, waxy solid, it improves texture, stability, and spreadability of creams, lotions, and ointments.
  • Waxy white solid at room temperature.
  • Acts as an emollient and thickening agent.
  • Functions as a nonionic emulsifier to stabilize oil-in-water systems.
  • Improves texture, spreadability, and product stability.
  • Suitable for topical pharmaceutical and cosmetic formulations.
  • High purity (99.1%).