Learn More
Apexbio Technology LLC Cetirizine,100mg CAS# 83881-51-0
SDP
Supplier:  Apexbio Technology LLC A3298100
Encompass_Preferred
Cetirizine, a second-generation antihistamine, is a major metabolite of hydroxyzine, and a racemic selective H1 receptor inverse agonist used in the treatment of allergies, hay fever, angioedema, and urticaria. Cetirizine crosses the blood-brain barrier only slightly, reducing the sedative side-effect common with older antihistamines. It has also been shown to inhibit eosinophil chemotaxis and LTB4 release. At a dosage of 20 mg, Boone et al. found that it inhibited the expression of VCAM-1 in patients with atopic dermatitis. The levorotary enantiomer of cetirizine, known as levocetirizine, is the more active form. From Wikipedia. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0623