Learn More
Medchemexpress LLC Cetirizine-d8 | 774596-22-4 | MFCD09840216 | 96.5% | 396.94 g/mol | C21H17D8ClN2O3 | 10 MG
SDP
Catalog No. 5000295018
Encompass_Preferred
Cetirizine-d8 is a deuterium-labeled analogue of cetirizine, a second-generation antihistamine. This isotopically labeled standard is supplied as a high-purity material for use in analytical applications, pharmacokinetic and metabolic studies, and other research workflows that require precise mass spectrometric quantitation.
  • High-purity deuterated reference material for analytical assays.
  • Suitable for LC-MS/MS internal standardization and method validation.
  • Facilitates accurate pharmacokinetic and metabolic studies.
  • Available as powder and in solvent for flexible preparation.
  • Provided in small stock sizes suitable for low-volume analytical work.
Catalog No. 50-002-95018 Supplier Medchemexpress LLC Supplier No. HY17042S110MG
Cetirizine-d8 is a deuterium-labeled analogue of cetirizine, a second-generation antihistamine. This isotopically labeled standard is supplied as a high-purity material for use in analytical applications, pharmacokinetic and metabolic studies, and other research workflows that require precise mass spectrometric quantitation.
  • High-purity deuterated reference material for analytical assays.
  • Suitable for LC-MS/MS internal standardization and method validation.
  • Facilitates accurate pharmacokinetic and metabolic studies.
  • Available as powder and in solvent for flexible preparation.
  • Provided in small stock sizes suitable for low-volume analytical work.