Learn More
Apexbio Technology LLC Cetirizine DiHCl 83881-52-1 100mg
SDP
Catalog No. 5000089926
Encompass_Preferred
Cetirizine dihydrochloride (CAS 83881-52-1) is a second-generation antihistamine structurally related to hydroxyzine It functions as a racemic selective inverse agonist of the histamine H1 receptor thereby attenuating histamine-mediated cellular responses Owing to its low penetration of the blood-brain barrier cetirizine exhibits reduced sedative effects compared to first-generation antihistamines This compound is widely utilized in biomedical research to investigate allergic pathways histaminergic signaling and the pharmacological modulation of conditions such as seasonal allergic rhinitis angioedema and urticaria
Catalog No. 50-000-89926 Supplier Apexbio Technology LLC Supplier No. B1547100
Cetirizine dihydrochloride (CAS 83881-52-1) is a second-generation antihistamine structurally related to hydroxyzine It functions as a racemic selective inverse agonist of the histamine H1 receptor thereby attenuating histamine-mediated cellular responses Owing to its low penetration of the blood-brain barrier cetirizine exhibits reduced sedative effects compared to first-generation antihistamines This compound is widely utilized in biomedical research to investigate allergic pathways histaminergic signaling and the pharmacological modulation of conditions such as seasonal allergic rhinitis angioedema and urticaria